KEY001146731 Key Organics / BIONET

5-chloro-1-[(2,6-dichlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140094
  • Cas Number : 339009-06-2
  • Molecular Weight : 332.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 288 - 289
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7E-405S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart