KEY001146760 Key Organics / BIONET

methyl 3-(3-chlorobenzamido)-6-methyl-2-oxo-2H-pyran-5-carboxylate

  • MDL number MFCD00139846
  • Cas Number : 339009-28-8
  • Molecular Weight : 321.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171 - 172
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84925

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7E-915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart