KEY001146763 Key Organics / BIONET

methyl 6-methyl-3-(4-methylbenzamido)-2-oxo-2H-pyran-5-carboxylate

  • MDL number MFCD00139849
  • Cas Number : 339009-31-3
  • Molecular Weight : 301.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 183
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84928

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7E-918_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart