KEY001146764 Key Organics / BIONET

ethyl 3-(4-methylbenzamido)-2-oxo-6-phenyl-2H-pyran-5-carboxylate

  • MDL number MFCD00139850
  • Cas Number : 339009-32-4
  • Molecular Weight : 377.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 186 - 188
Iupac Name ethyl 3-[(4-methylbenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate
Inchi Code 1S/C22H19NO5/c1-3-27-21(25)17-13-18(23-20(24)16-11-9-14(2)10-12-16)22(26)28-19(17)15-7-5-4-6-8-15/h4-13H,3H2,1-2H3,(H,23,24)
Inchi Key QTUICLYEVNXSIY-UHFFFAOYSA-N

Description

Melting Point 186 - 188
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84929

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7E-919_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart