KEY001146778 Key Organics / BIONET

methyl 3-[(4-chlorophenyl)methoxy]thiophene-2-carboxylate

  • MDL number MFCD00173481
  • Cas Number : 339009-41-5
  • Molecular Weight : 282.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 104 - 106
Iupac Name methyl 3-[(4-chlorobenzyl)oxy]-2-thiophenecarboxylate
Inchi Code 1S/C13H11ClO3S/c1-16-13(15)12-11(6-7-18-12)17-8-9-2-4-10(14)5-3-9/h2-7H,8H2,1H3
Inchi Key DUVIRIQWBNKRPB-UHFFFAOYSA-N

Description

Melting Point 104 - 106
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84950

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7F-011_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart