KEY001146885 Key Organics / BIONET

methyl 3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate

  • MDL number MFCD00232259
  • Cas Number : 339010-38-7
  • Molecular Weight : 308.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85082

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7G-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart