KEY001146922 Key Organics / BIONET

4-(4-chlorophenyl)-1H-pyrazol-5-amine

  • MDL number MFCD00231866
  • Cas Number : 40545-65-1
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63 - 65
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Low Melting Solid,Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85121

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7G-047_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart