KEY001147026 Key Organics / BIONET

methyl 4-{[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)oxy]methyl}benzoate

  • MDL number MFCD00172629
  • Cas Number : 339011-73-3
  • Molecular Weight : 311.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 172 - 174
Iupac Name methyl 4-{[(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)oxy]methyl}benzoate
Inchi Code 1S/C17H13NO5/c1-22-17(21)12-8-6-11(7-9-12)10-23-18-15(19)13-4-2-3-5-14(13)16(18)20/h2-9H,10H2,1H3
Inchi Key WDWPUOCISOEHHR-UHFFFAOYSA-N

Description

Melting Point 172 - 174
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85280

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7G-934_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart