KEY001147206 Key Organics / BIONET

5-({[(2,6-dichlorophenyl)methyl]sulfanyl}methyl)-4-ethyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00244410
  • Cas Number : 344269-70-1
  • Molecular Weight : 334.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 178
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85470

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7H-439S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart