KEY001148252 Key Organics / BIONET

5-({[(2,4-dichlorophenyl)methyl]sulfanyl}methyl)-4-(propan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00244456
  • Cas Number : 344272-66-8
  • Molecular Weight : 348.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 137 - 140
Iupac Name 5-{[(2,4-dichlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazole-3-thiol
Inchi Code 1S/C13H15Cl2N3S2/c1-8(2)18-12(16-17-13(18)19)7-20-6-9-3-4-10(14)5-11(9)15/h3-5,8H,6-7H2,1-2H3,(H,17,19)
Inchi Key HOVAMNOLUNEIKI-UHFFFAOYSA-N

Description

Melting Point 137 - 140
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85519

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7H-495S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart