KEY001148295 Key Organics / BIONET

1-(3,4-dichlorophenyl)-2-methanesulfonylethan-1-one

  • MDL number MFCD00202208
  • Cas Number : 344275-49-6
  • Molecular Weight : 267.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85565

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7H-572S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart