KEY001148322 Key Organics / BIONET

3-(4-methylphenyl)-2,1-benzoxazole-5-carbaldehyde

  • MDL number MFCD00663843
  • Cas Number : 328547-38-2
  • Molecular Weight : 237.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 154
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85593

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7H-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart