KEY001148326 Key Organics / BIONET

(2E)-3-[3-(4-methylphenyl)-2,1-benzoxazol-5-yl]prop-2-enoic acid

  • MDL number MFCD00663847
  • Cas Number : 344276-77-3
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 295
Iupac Name (2E)-3-[3-(4-methylphenyl)-2,1-benzisoxazol-5-yl]-2-propenoic acid
Inchi Code 1S/C17H13NO3/c1-11-2-6-13(7-3-11)17-14-10-12(5-9-16(19)20)4-8-15(14)18-21-17/h2-10H,1H3,(H,19,20)/b9-5+
Inchi Key YPDPEHKRYOMXIP-WEVVVXLNSA-N

Description

Melting Point 295
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85597

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7H-928_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart