KEY001148388 Key Organics / BIONET

2-chloro-1-(4-hydroxy-3,5-dimethylphenyl)ethan-1-one

  • MDL number MFCD00232525
  • Cas Number : 40943-25-7
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 111
Purity 95
Storage temp. RT
Physical Form White to Red Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85670

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7J-328S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart