KEY001148420 Key Organics / BIONET

4-(4-chlorophenyl)-2-(4-methylphenyl)-4-oxobutanoic acid

  • MDL number MFCD00664382
  • Cas Number : 70596-90-6
  • Molecular Weight : 302.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 188
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85710

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-525S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart