KEY001148442 Key Organics / BIONET

4-(4-chlorophenyl)-2-(3-fluorophenyl)-4-oxobutanoic acid

  • MDL number MFCD00664404
  • Cas Number : 344280-97-3
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 164
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85732

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-557S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart