KEY001148444 Key Organics / BIONET

2-(4-chlorophenyl)-4-oxo-4-phenylbutanoic acid

  • MDL number MFCD00664406
  • Cas Number : 39206-70-7
  • Molecular Weight : 288.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85734

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-560S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart