KEY001148455 Key Organics / BIONET

4-(4-chlorophenyl)-4-oxo-2-[4-(propan-2-yl)phenyl]butanoic acid

  • MDL number MFCD00664417
  • Cas Number : 344281-36-3
  • Molecular Weight : 330.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 157
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85746

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-579S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart