KEY001148463 Key Organics / BIONET

5-(4-chlorophenyl)pyrimidine-2-thiol

  • MDL number MFCD00232539
  • Cas Number : 31408-24-9
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 265 - 268
Iupac Name 5-(4-chlorophenyl)-2(3H)-pyrimidinethione
Inchi Code 1S/C10H7ClN2S/c11-9-3-1-7(2-4-9)8-5-12-10(14)13-6-8/h1-6H,(H,12,13,14)
Inchi Key TWTICSLSIMZTBQ-UHFFFAOYSA-N

Description

Melting Point 265 - 268
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85754

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7J-591S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart