KEY001148483 Key Organics / BIONET

2-(methylsulfanyl)-5-phenylpyrimidine

  • MDL number MFCD00204407
  • Cas Number : 31408-20-5
  • Molecular Weight : 202.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-611S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart