KEY001148519 Key Organics / BIONET

5-[3-(trifluoromethyl)phenyl]pyrimidine-2-thiol

  • MDL number MFCD00243479
  • Cas Number : 344282-81-1
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262 - 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85811

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-666S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart