KEY001148659 Key Organics / BIONET

{[4-(methoxycarbonyl)phenyl]methyl}triphenylphosphanium bromide

  • MDL number MFCD00082765
  • Cas Number : 1253-46-9
  • Molecular Weight : 491.365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255 - 257
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85992

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7K-902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart