KEY001148661 Key Organics / BIONET

2-amino-1-(naphthalen-2-yl)ethan-1-ol

  • MDL number MFCD00974251
  • Cas Number : 5696-74-2
  • Molecular Weight : 187.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 124
Purity 95
Storage temp. Store under N2
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85994

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7K-904_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart