KEY001148671 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(naphthalen-2-yl)prop-2-en-1-one

  • MDL number MFCD00179142
  • Cas Number : 138716-23-1
  • Molecular Weight : 225.291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86006

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7L-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart