KEY001148682 Key Organics / BIONET

2-[5-(2,4-dichlorobenzoyl)thiophen-2-yl]acetonitrile

  • MDL number MFCD00664601
  • Cas Number : 339014-61-8
  • Molecular Weight : 296.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86019

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7L-017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart