KEY001148880 Key Organics / BIONET

2-{1-[(4-fluorophenyl)methyl]-1H-indol-3-yl}acetic acid

  • MDL number MFCD00794686
  • Cas Number : 176204-51-6
  • Molecular Weight : 283.302

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 166
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86243

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7L-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart