KEY001148881 Key Organics / BIONET

2-[1-(6-chloropyridazin-3-yl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD00794687
  • Cas Number : 339016-30-7
  • Molecular Weight : 287.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 220 - 222
Iupac Name [1-(6-chloro-3-pyridazinyl)-1H-indol-3-yl]acetic acid
Inchi Code 1S/C14H10ClN3O2/c15-12-5-6-13(17-16-12)18-8-9(7-14(19)20)10-3-1-2-4-11(10)18/h1-6,8H,7H2,(H,19,20)
Inchi Key IDHJLSIRISGRPI-UHFFFAOYSA-N

Description

Melting Point 220 - 222
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86244

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7L-906_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart