KEY001148884 Key Organics / BIONET

2-{1-[(3-chlorophenyl)methyl]-1H-indol-3-yl}acetic acid

  • MDL number MFCD00794689
  • Cas Number : 339016-33-0
  • Molecular Weight : 299.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 105 - 107
Iupac Name [1-(3-chlorobenzyl)-1H-indol-3-yl]acetic acid
Inchi Code 1S/C17H14ClNO2/c18-14-5-3-4-12(8-14)10-19-11-13(9-17(20)21)15-6-1-2-7-16(15)19/h1-8,11H,9-10H2,(H,20,21)
Inchi Key BMZCMTIGPMYJKO-UHFFFAOYSA-N

Description

Melting Point 105 - 107
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86247

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7L-909_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart