KEY001148900 Key Organics / BIONET

methyl 4-[(pyrimidin-2-yl)amino]butanoate

  • MDL number MFCD00794701
  • Cas Number : 339016-47-6
  • Molecular Weight : 195.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86264

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7L-927_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart