KEY001148922 Key Organics / BIONET

3-(thiophen-2-yl)-2,5-dihydrofuran-2,5-dione

  • MDL number MFCD00974740
  • Cas Number : 339016-64-7
  • Molecular Weight : 180.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 128
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7M-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart