KEY001149120 Key Organics / BIONET

[(4-methylphenyl)methyl]urea

  • MDL number MFCD01314625
  • Cas Number : 54582-34-2
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 198 - 199
Iupac Name N-(4-methylbenzyl)urea
Inchi Code 1S/C9H12N2O/c1-7-2-4-8(5-3-7)6-11-9(10)12/h2-5H,6H2,1H3,(H3,10,11,12)
Inchi Key CWQOSUOSPHUYTM-UHFFFAOYSA-N

Description

Melting Point 198 - 199
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7M-748_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart