KEY001150218 Key Organics / BIONET

methyl 5-cyano-1-(4-fluorophenyl)-4-methyl-6-oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD01315976
  • Cas Number : 339018-13-2
  • Molecular Weight : 287.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 163 - 165
Iupac Name methyl 5-cyano-1-(4-fluorophenyl)-4-methyl-6-oxo-1,6-dihydro-3-pyridazinecarboxylate
Inchi Code 1S/C14H10FN3O3/c1-8-11(7-16)13(19)18(17-12(8)14(20)21-2)10-5-3-9(15)4-6-10/h3-6H,1-2H3
Inchi Key LQOZAFGJZXPNLQ-UHFFFAOYSA-N

Description

Melting Point 163 - 165
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86612

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7M-931_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart