KEY001150252 Key Organics / BIONET

4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzonitrile

  • MDL number MFCD01443858
  • Cas Number : 339018-30-3
  • Molecular Weight : 250.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 134 - 136
Iupac Name 4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzonitrile
Inchi Code 1S/C11H7ClN2OS/c12-11-14-6-10(16-11)7-15-9-3-1-8(5-13)2-4-9/h1-4,6H,7H2
Inchi Key DXWMIWQOLXLUMR-UHFFFAOYSA-N

Description

Melting Point 134 - 136
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86646

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7N-029_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart