KEY001150616 Key Organics / BIONET

2-(4-bromophenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD01067251
  • Cas Number : 294866-41-4
  • Molecular Weight : 288.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 175 - 177
Iupac Name 2-(4-bromophenyl)-1,3-thiazolidine-4-carboxylic acid
Inchi Code 1S/C10H10BrNO2S/c11-7-3-1-6(2-4-7)9-12-8(5-15-9)10(13)14/h1-4,8-9,12H,5H2,(H,13,14)
Inchi Key TYLJHTCYCWHWRG-UHFFFAOYSA-N

Description

Melting Point 175 - 177
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87035

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7P-024_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart