KEY001150633 Key Organics / BIONET

(2E)-3-amino-3-(4-bromophenyl)prop-2-enenitrile

  • MDL number MFCD01871637
  • Cas Number : 748807-82-1
  • Molecular Weight : 223.073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87057

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7P-061_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart