KEY001150644 Key Organics / BIONET

1,1,1-trifluoro-3-({[3-(trifluoromethyl)phenyl]methyl}amino)propan-2-ol

  • MDL number MFCD01871640
  • Cas Number : 478068-12-1
  • Molecular Weight : 287.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87069

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7P-085_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart