KEY001150941 Key Organics / BIONET

(2E)-3-(3-methylthiophen-2-yl)prop-2-enoic acid

  • MDL number MFCD00209366
  • Cas Number : 77741-66-3
  • Molecular Weight : 168.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161 - 164
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-0651_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart