KEY001151307 Key Organics / BIONET

5-chloro-1-[(4-chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140420
  • Cas Number : 339024-38-3
  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 230.5 - 231.5
Iupac Name 5-chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarboxylic acid
Inchi Code 1S/C13H9Cl2NO3/c14-10-3-1-8(2-4-10)6-16-7-9(13(18)19)5-11(15)12(16)17/h1-5,7H,6H2,(H,18,19)
Inchi Key IQZOWCJHDNIYJO-UHFFFAOYSA-N

Description

Melting Point 230.5 - 231.5
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88607

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8E-351S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart