KEY001151326 Key Organics / BIONET

5-chloro-1-[(3-chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140433
  • Cas Number : 339024-74-7
  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 229 - 230.5
Iupac Name 5-chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarboxylic acid
Inchi Code 1S/C13H9Cl2NO3/c14-10-3-1-2-8(4-10)6-16-7-9(13(18)19)5-11(15)12(16)17/h1-5,7H,6H2,(H,18,19)
Inchi Key RZLVKMJJANTOPJ-UHFFFAOYSA-N

Description

Melting Point 229 - 230.5
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88629

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8E-393S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart