KEY001151528 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-one

  • MDL number MFCD00231641
  • Cas Number : 339029-35-5
  • Molecular Weight : 278.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62 - 65
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88853

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8F-941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart