KEY001151538 Key Organics / BIONET

1-(3-chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD00141204
  • Cas Number : 339029-56-0
  • Molecular Weight : 266.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 271 - 273
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8G-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart