KEY001151670 Key Organics / BIONET

(2E)-3-[(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD00202686
  • Cas Number : 339096-60-5
  • Molecular Weight : 337.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89038

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8G-954_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart