KEY001151689 Key Organics / BIONET

methyl 3-[(3-oxobutan-2-yl)oxy]thiophene-2-carboxylate

  • MDL number MFCD00173639
  • Cas Number : 339096-81-0
  • Molecular Weight : 228.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54 - 56
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89059

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8H-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart