KEY001151844 Key Organics / BIONET

2-{1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indol-3-yl}acetohydrazide

  • MDL number MFCD00664547
  • Cas Number : 339099-27-3
  • Molecular Weight : 368.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 153
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89232

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8H-950_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart