KEY001152081 Key Organics / BIONET

2-{[(4-chlorophenyl)methyl]sulfanyl}benzoic acid

  • MDL number MFCD01315364
  • Cas Number : 15887-84-0
  • Molecular Weight : 278.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212 - 215
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89497

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8K-511S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart