KEY001152092 Key Organics / BIONET

ethyl 1-(4-phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00665285
  • Cas Number : 159885-61-7
  • Molecular Weight : 367.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89510

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8K-533S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart