KEY001152146 Key Organics / BIONET

2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)-3-phenylpropanoic acid

  • MDL number MFCD00664644
  • Cas Number : 96017-10-6
  • Molecular Weight : 281.311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212 - 214
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89572

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8L-018_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart