KEY001152203 Key Organics / BIONET

4-[5-(4-methoxyphenyl)-1H-pyrazol-3-yl]piperidine

  • MDL number MFCD00665098
  • Cas Number : 103660-47-5
  • Molecular Weight : 257.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 183
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89656

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8L-707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart