KEY001153259 Key Organics / BIONET

5-{[(2-chlorophenyl)formamido]methyl}-4,5-dihydro-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD01315982
  • Cas Number : 339101-05-2
  • Molecular Weight : 282.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8L-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart