KEY001153277 Key Organics / BIONET

4-(methoxycarbonyl)pyridine-3-carboxylic acid

  • MDL number MFCD00111513
  • Cas Number : 24202-74-2
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 183 - 185
Iupac Name 4-(methoxycarbonyl)nicotinic acid
Inchi Code 1S/C8H7NO4/c1-13-8(12)5-2-3-9-4-6(5)7(10)11/h2-4H,1H3,(H,10,11)
Inchi Key QLOYEIURVSRKOV-UHFFFAOYSA-N

Description

Melting Point 183 - 185
Purity 99%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89721

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8L-924_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart